missing translation for 'onlineSavingsMsg'
Learn More

Adenosine, MP Biomedicals

Product Code. 11421145 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421145 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421145 Supplier MP Biomedicals Supplier No. 0219980380
  • ONLINE EXCLUSIVE PRICE
    310.00 €
    294.00 € / 100g
    Save 16.00 €
    5% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Adenosine is used in topical treatment of foot wounds in diabetes mellitus and also used for the treatment or prevention of cardiac arrhythmias (irregular heartbeat)
  • It is used to cause dilation of the blood vessels

Specifications

CAS 58-61-7
Melting Point 233°C to 238°C
Color White
Molecular Formula C10H13N5O4
Linear Formula C10H13N5O4
MDL Number MFCD00005752
Quantity 100 g
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 267.25
PubChem CID 60961
ChEBI CHEBI:16335
Formula Weight 267.2
Grade USP
Physical Form Powder
Chemical Name or Material ADENOSINE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.