missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Adhesive Labels
Click to view available options
Color:
Black
Blue
Green
Orange
Display Text:
Chloroform
Dist. Water
Ethanol
Ethyl Acetate
Methanol
Methylene Cloride
Unit Size:
Pack of 50
Description
- For use with wash bottles and plastic or glass reagent bottles, as recommended by the NFPA (National Fire Protection Agency)
- Complete with risk assessment numbers, recommendations for use and special notices shown in reagent catalogues
Specifications
Specifications
| Description | Transparent with colored text |
| Color | Orange |
| Display Text | Ethyl Acetate |
| Quantity | 50/Pk. |
| Dimensions (LxW) | 130 x 35 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction