missing translation for 'onlineSavingsMsg'
Learn More

Adipic acid, 99%

Product Code. 10132320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
2.5kg
25kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10132320 500 g Plastic bottle 500g
10090550 1 kg Plastic bottle 1kg
10538280 2.5 kg Plastic bottle 2.5kg
10454745 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10132320 Supplier Thermo Scientific Acros Supplier No. 102815000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-04-9 | C6H10O4 | 146.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-04-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00004420
InChI Key WNLRTRBMVRJNCN-UHFFFAOYSA-N
Synonym adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
PubChem CID 196
ChEBI CHEBI:30832
IUPAC Name hexanedioic acid
SMILES C(CCC(=O)O)CC(=O)O

Specifications

Melting Point 151.0°C to 153.0°C
Boiling Point 337.0°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Quantity 500 g
Flash Point 196°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 99%
Linear Formula HO2C(CH2)4CO2H
Beilstein 02, 649
Fieser 01,15
Merck Index 15, 150
Formula Weight 146.14
Percent Purity 99%
Chemical Name or Material Adipic acid
Show More Show Less
compliance-icons
Product Identifier
  • Adipic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.