missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic acid, 99%
Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
missing translation for 'unitSize'
1kg
2.5kg
25kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 151.0°C to 153.0°C |
| Boiling Point | 337.0°C |
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Color | White |
| Assay Percent Range | 99% |
| Linear Formula | HO2C(CH2)4CO2H |
| Beilstein | 02, 649 |
| Fieser | 01,15 |
| Merck Index | 15, 150 |
| Show More |
Safety and Handling
Product Identifier
- Adipic acid
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction