missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid Dihydrazide, TRC

Product Code. 17920625
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920625 50 g 50g
17981335 25 g 25g
17981765 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920625 Supplier Toronto Research Chemicals Supplier No. TRCA29159550G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6 H14 N4 O2
Molecular Weight (g/mol) 174.2
Synonym Hexanedioic Acid Dihydrazide, ADH, ADH (hydrazide), ADH 4S, ADH-J, ADH-S, Adipic Dihydrazide, Adipoyl Dihydrazide, Adipoyl Hydrazide, Adipoyldihydrazine, Ajicure ADH, BK 1000Z, BT 1000Z, NSC 29542, NSC 3378, Qualimer ADH, T 2210, Adipohydrazide
IUPAC Name hexanedihydrazide
SMILES NNC(CCCCC(NN)=O)=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H14N4O2/c7-9-5(11)3-1-2-4-6(12)10-8/h1-4,7-8H2,(H,9,11)(H,10,12)
Chemical Name or Material Adipic Acid Dihydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.