missing translation for 'onlineSavingsMsg'
Learn More

Adipic dihydrazide, 97%

Product Code. 11482078
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482078 500 g 500g
11462078 25 g 25g
11472078 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482078 Supplier Thermo Scientific Alfa Aesar Supplier No. A15119.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1071-93-8 | C6H14N4O2 | 174.20 g/mol

Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.

Solubility
Soluble in water, alcohol and acetone.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00007614
InChI Key IBVAQQYNSHJXBV-UHFFFAOYSA-N
Synonym adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
PubChem CID 66117
IUPAC Name hexanedihydrazide
SMILES NNC(=O)CCCCC(=O)NN

Specifications

Melting Point 180°C to 182°C
Odor Odorless
Quantity 500 g
Beilstein 973863
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material Adipic dihydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.