missing translation for 'onlineSavingsMsg'
Learn More

Adipic semialdehyde methyl ester, 95%

Product Code. 11985111 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 11985111

Brand: Thermo Scientific Acros 438912500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6654-36-0 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6654-36-0
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Boiling Point 59°C to 61°C (1.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C7H12O3
Linear Formula OHCCH2CH2CH2CH2CO2CH3
Quantity 250 mg
Synonym adipic semialdehyde methyl ester, methyl 5-formylvalerate, methyl adipaldehydate, 6-oxo-hexanoic acid methyl ester, 6-oxohexanoic acid methyl ester, hexanoic acid, 6-oxo-, methyl ester, 5-carbomethoxy-1-pentanal, hexanoic acid,6-oxo-,methyl ester, methyl 6-oxo-hexanoate, methyl 5-formylpentanoate
InChI Key FDNFXHCDOASWAY-UHFFFAOYSA-N
SMILES COC(=O)CCCCC=O
IUPAC Name methyl 6-oxohexanoate
Molecular Weight (g/mol) 144.17
PubChem CID 81164
Formula Weight 144.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Adipic semialdehyde methyl ester
Show More Show Less
compliance-icons
Product Identifier
  • Adipic semialdehyde methyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.