missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AdipoRon hydrochloride

Product Code. 30002052
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002052 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30002052 Supplier R&D Systems™ Supplier No. 5096/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Orally active AdipoR1 and AdipoR2 agonist; antidiabetic

Agonist of adiponectin receptors AdipoR1 and AdipoR2 (Kd values are 1.8 and 3.1 μM respectively). Activates AMPK and PPARα signaling; ameliorates insulin resistance, dyslipidemia and glucose intolerance in db/db mice. Orally active antidiabetic agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1781835-20-8
Quantity 50 mg
Product Line Tocris
Molecular Formula C27H29ClN2O3
Formula Weight 464.98
Molecular Weight (g/mol) 464.99
Chemical Name or Material AdipoRon hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.