missing translation for 'onlineSavingsMsg'
Learn More

Adipoyl chloride, 98%

Product Code. 11454554
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454554 25 g 25g
11464554 100 g 100g
11474554 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454554 Supplier Thermo Scientific Alfa Aesar Supplier No. A13168.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-50-2 | C6H8Cl2O2 | 183.028 g/mol

Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

Solubility
Immiscible with water.

Notes
Incompatible strong oxidizing agents, bases and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-50-2
Molecular Formula C6H8Cl2O2
Molecular Weight (g/mol) 183.028
MDL Number MFCD00000759
InChI Key PWAXUOGZOSVGBO-UHFFFAOYSA-N
Synonym adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242
PubChem CID 61034
IUPAC Name hexanedioyl dichloride
SMILES C(CCC(=O)Cl)CC(=O)Cl

Specifications

Density 1.255
Boiling Point 105°C to 107°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Linear Formula ClCO(CH2)4COCl
Refractive Index 1.4705
Quantity 25 g
UN Number UN3265
Beilstein 507709
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 183.03
Percent Purity 98%
Chemical Name or Material Adipoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Adipoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Adipoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.