missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Benches, Rolling Carts and Bases for Erlab Enclosures: Erlab benches and rolling carts are designed to supPort ductless enclosures and filtering cabinets at the correct working height, in complete safety. Each model is dimensioned for a specific enclosure reference to guarantee stability and a perfect match with the equipment footprint.
- Stability and Ergonomics:
- Robust steel structure sized for the load of the corresponding enclosure
- Chemically resistant, easy-to-clean finish
- Fixed, mobile, levelling-feet, mechanically adjustable and electrically adjustable versions available
- Height-adjustable models allow the working position to be adapted to each operator, seated or standing
- Lockable castors on rolling versions for safe repositioning of the workstation
- Flexible Laboratory Layout: Rolling carts make it possible to move a complete filtration workstation from one room to another as projects evolve, while fixed benches provide a permanent, space-efficient installation. Optional accessories such as retractable shelves and footrests further improve daily comfort and storage capacity.
- Key Benefits:
- Dedicated supPort designed for a specific Erlab enclosure.
- Stable, robust structure with chemically resistant finish.
- Fixed, mobile and height-adjustable versions available.
- Improved operator ergonomics and workstation flexibility.
- Simple assembly and easy maintenance.
Specifications
Specifications
| Product Type | Adjustable Base with Wheels |
| For Use With (Equipment) | Solis, Olis, Lois 1800 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?