Learn More
Leica Microsystems Adjustable, Low-eyepoint Eyepiece, E-Series
Click to view available options
Magnification Power:
20X
Unit Size:
Each
Description
- Adjustable.
- Low eyepoint.
Specifications
Specifications
| Type | Adjustable Low-eyepoint Microscope Eyepiece |
| Field of View | 12 |
| For Use With (Equipment) | E-Series Educational StereoZoom™ Microscopes |
| Magnification Power | 20X |
| Quantity | 1 Ea. |