missing translation for 'onlineSavingsMsg'
Learn More

ADK Stab NA 21E, TRC

Product Code. 30011517
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011517 1 g 1g
30010328 250 mg 250mg
30007560 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011517 Supplier Toronto Research Chemicals Supplier No. TRCA2918101G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151841-65-5
Molecular Formula C58H85AlO9P2
Molecular Weight (g/mol) 1015.22
Synonym Hydroxybis[2,4,8,10-tetrakis(1,1-dimethylethyl)-6-(hydroxy-κO)-12H-dibenzo[d,g][1,3,2]dioxaphosphocin 6-oxidato]aluminum, 2,2-Methylenebis(4,6-di-tert-butylphenyl)phosphate Aluminum Hydroxide Salt, Aluminium Hydroxy bis[2,2'-Methylene-bis(4,6-di-tert-butylphenyl) phosphate], Aluminum Hydroxybis[2,2'-methylenebis[4,6-di(tert-butyl)phenyl]phosphate], Aluminum Methylenebis(2,4-di-tert-butyl-benzyloxy)phosphate, Bis(2,4,8,10-tetra-tert-butyl-6-hydroxy-12H-dibenzo[d,g][1,2,3]dioxaphosphocin-6-oxide) Aluminum Hydroxide Salt, Bis(2,4,8,10-tetra-tert-butyl-6-hydroxy-12H-dibenzo[d,g][1,2,3]dioxaphosphocin-6-oxide)aluminum Hydroxide, Bis(2,4,8,10-tetra-tert-butyl-6-hydroxy-12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-oxide) Aluminum Hydroxide, Bis[2,2'-methylenebis(4,6-di-tert-butylphenyl) phosphate] Aluminum Hydroxide Salt, HBP, Hydroxyaluminum-bis[2,2-methylene-bis(4,6-di-tert-butylphenyl)phosphate], NA 13, NA 21E, TMP 5;
IUPAC Name aluminum;1,3,7,9-tetratert-butyl-11-oxido-5H-benzo[d][1,3,2]benzodioxaphosphocine 11-oxide;hydroxide
SMILES O=P1(OC2=C(C(C)(C)C)C=C(C(C)(C)C)C=C2CC3=CC(C(C)(C)C)=CC(C(C)(C)C)=C3O1)O[Al](O)OP4(OC5=C(C(C)(C)C)C=C(C(C)(C)C)C=C5CC6=CC(C(C)(C)C)=CC(C(C)(C)C)=C6O4)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C29H43O4P.Al.H2O/c2*1-26(2,3)20-14-18-13-19-15-21(27(4,5)6)17-23(29(10,11)12)25(19)33-34(30,31)32-24(18)22(16-20)28(7,8)9;;/h2*14-17H,13H2,1-12H3,(H,30,31);;1H2/q;;+3;/p-3
Formula Weight 1014.5484
Chemical Name or Material ADK Stab NA 21E
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.