Learn More
Vilbar Lourmat BIO1D SOFTWARE
Product Code. 11761159
Change view
Click to view available options
Product Type:
Administrator Software CFR21
Unit Size:
Each
Product Code. Product Type Unit Size
11761159 Administrator Software CFR21 Each
1 options
Product Code. 11761159 Supplier Vilbar Lourmat Supplier No. 141010011
  • ONLINE EXCLUSIVE PRICE
    1625.00 €
    1384.39 € / Pack of 1
    Save 240.61 €
    14% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Administrator software ideal for calculating automatic band detection and molecular weight.
  • Pentium, 3.2 GHz, FSB 800 MHz, 33 MHz PCI bus, 512 MB Ram, 10 GB hard disk with at least 100 MB of free space for software installation and image backup, 1280 x 1024 VGA card in 16 million colors (24 Bits), and at least 1 PCI port and 1 free USB port
  • Bio1-D software
  • Transformation of the 1D image into a 3D graph for quantification of the bands
  • Automatic band detection and molecular weight calculation
  • Simultaneous display of the gel and the values ​​of each strip on the same screen
  • Recalculation of values ​​according to a reference calibration curve
  • Quantitative analysis of microplates
  • Colony count according to their characteristics (volume, area, perimeter)
  • Ieporting integrating images of gels, result tables, curves that can be printed or exported in a Windows ™ compatible format
  • Full GLP compliance
Specifications
Product Type Administrator Software CFR21