missing translation for 'onlineSavingsMsg'
Learn More

Clinitrol™ 290 Reference Solution; Advanced Instruments

Product Code. 13214750
Encompass
Change view
Click to view available options
For Use With (Equipment):
3D3, 3250 and 3900 osmometers
Quantity:
10 x 5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Quantity Unit Size
13214750 3D3, 3250 and 3900 osmometers 10 x 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13214750 Supplier Brand Not Defined Supplier No. 3LA029

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Critical normal human serum midpoint of 290mOsm

  • Checks performance of freezing-point osmometers in the critical area of normal serum values
  • Contains known and controlled quantities of some of the same solutes which comprise osmotically active particles of serum
  • Pipetted into a tube and run just like normal serum
  • Runs as the first and last samples in a series of tests to monitor operator technique and instrument performance
  • Does not require mixing, reconstituting or refrigerating; one-year shelf life

Specifications

Certifications/Compliance CE
Color Clear
Content And Storage 2°C to 30°C
Concentration 290 mOsm/kg H2O
For Use With (Equipment) 3D3, 3250 and 3900 osmometers
Sample Type Serum
Type Reference Solution
Includes Sodium Chloride (NaCl) and 2-Hydroxybiphenyl Sodium Salt Tetrahydrate
Intended Use Osmometer instrument calibration and quality control
Quantity 10 x 5 mL
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.