Learn More
Advanced Instruments Osmometer Disposables
Product Code. 15118596
Encompass
Change view
Click to view available options
For Use With (Equipment):
3D3 and 3250 Osmometer
A2O Advanced Automated Osmometer
Advanced Model 2020 Osmometer
Product Type:
Air Filter
Micro-Sample Tube
Sample Tube
Unit Size:
Each
Pack of 500
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Product Type Unit Size
15118596 Advanced Model 2020 Osmometer Sample Tube Pack of 500
13400547 3D3 and 3250 Osmometer Air Filter Pack of 6
15128606 3D3 and 3250 Osmometer Air Filter Each
15745672 A2O Advanced Automated Osmometer Micro-Sample Tube Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15118596 Supplier Advanced Instruments Supplier No. 202825
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Disposables for use with Advanced osmometers
A wide assortment of accessories are available for use with Advanced Instruments Osmometers, including air filters, sample tubes, pipets, cleaning wipes and more.
Specifications
Product Type Sample Tube
For Use With (Equipment) Advanced Model 2020 Osmometer
Disposable Single-use