missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Adventurer™ Semi-micro Balance

Product Code. 17901292 Shop All Ohaus Products
Change view
Click to view available options
Model:
AX125D
AX125DM
AX225D
AX225DM
AX85
AX85M
Capacity:
102 g/220 g
82 g
82 g/120 g
Readability:
0.00001 g
0.00001 g, 0.0001 g
Unit Size:
1 unit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Capacity Readability unitSize
17901292 AX85 82 g 0.00001 g 1 unit
17981292 AX85M 82 g 0.00001 g 1 unit
17911292 AX125D 82 g/120 g 0.00001 g, 0.0001 g 1 unit
17921292 AX125DM 82 g/120 g 0.00001 g, 0.0001 g 1 unit
17951302 AX225D 102 g/220 g 0.00001 g, 0.0001 g 1 unit
17971292 AX225DM 102 g/220 g 0.00001 g, 0.0001 g 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17901292 Supplier OHAUS™ Supplier No. 30852308
  • ONLINE EXCLUSIVE PRICE
    5886.24 €
    5235.13 € / 1 units
    Save 651.11 €
    11% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Intuitive Adventurer™ Semi-micro Balance designed for routine weighing.

Intuitive Balances Designed for Routine Weighing:

Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, three level user management to fulfill GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.

Unique Features Include:

  • Adventurer balances feature a color touchscreen, icon-based user interface, and an ergonomic design -making them easy to configure and use.
  • Features such as specialized weighing modes, multiple connectivity options, and AutoCal™ provide versatility and flexibility for a variety of applications.
  • Durable construction, large weighing surfaces, a space-saving draftshield design, and full housing in-use cover allow for use in lab, education and industrial environments.

Stability, Accuracy, and Fast Operation Ensure Optimal Weighing Results in Routine Weighing Tasks:

Weighing Performance:

  • Delivers stable and reliable weighing results for routine weighing tasks.

Stabilization Time:

  • Adventurer's fast stabilization time improves productivity in the laboratory.

Calibration:

  • AutoCal™ — Selected models feature OHAUS' automatic internal calibration system that performs routine maintenance by calibrating the balance daily.
  • External Calibration — Traditional calibration in which the operator manually calibrates the balance with their choice of calibration weight value to ensure accuracy available on every model.

Color Touchscreen Offers Easy and Fast Operation of Adventurer's Applications

  • Operate and access Adventurer's nine application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen.
  • Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves.
  • In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print.

Equipped with the Connectivity and Functional Features Required in Laboratories

Dual USB Ports:

  • A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance.
  • A second USB device is located at the rear of the balance that can be used to connect the balance to a PC.
  • The connectivity options help meet traceability requirements in traditional installations.

Label Printing Function:

  • Easy to link with Zebra printer and have one built-in label printing template.

Balance Profiles:

  • The cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances.

Below Minimum Sample Weight Indication:

  • When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit. Simply increase your sample weight to assure that your results are up to your standard.

Space-saving Draftshield Designed to Improve User Experience and Accessibility:

  • Draftshield doors are constructed of two glass panels, reducing the space required on the lab bench when the doors are open.
  • Wide door entry provides unobstructed access and allows larger weighing vessels to be easily placed on the pan, reducing the chance of accidental spillage.
  • Easy to keep clean in order to ensure a safe workspace by minimizing contamination.

Power Saving Functions:

  • The Adventurer is designed with various power-saving features that help reduce the environmental impact.
  • These include auto-off and brightness controls.

User Management Function:

  • Ensures data security and data traceability.
  • 3 level user management function ensures data security and data traceability requirements.
  • One administrator, two supervisors and 10 users have preset accessibility in the software.

Real Time Clock with GLP/GMP Data:

  • A real-time clock function keeps accurate time even during power loss.
  • GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements.
TRUSTED_SUSTAINABILITY

Specifications

Calibration Internal
Certifications/Compliance Metrology: OIML R76, EN 45501 (Class I, nmax 320000; Class II, nmax 52000), Product Safety: EN 61010-1, IEC 61010-1Electromagnetic Compatibility: IEC 61326-1, EN 61326-1 (emissions Class B, immunity Industrialrequirements) GLP/GMP Compliant
Connection Type RS232 (Included), USB, USB Host
Capacity 82 g
Display Type Color Graphic
Readability 0.00001 g
Includes In-use cover
Linearity 0.1 mg
Min. Sample Weight 20.0 mg
Model AX85
Repeatability 0.00002 g
Stabilization Time 8.0 sec.
Type Semi-Micro Balance
Dimensions (L x W x H) 354 x 230 x 340 mm
Length (English) 13.9 in.
Length (Metric) 354.0 mm
Width (English) 9.1 in.
Width (Metric) 230.0 mm
Height (English) 13.4 in.
Height (Metric) 340.0 mm
Diameter (English) Outer Pan 3.1 in.
Diameter (Metric) Outer Pan 80.0 mm
Weight (English) 11.2 lb.
Weight (Metric) 5.1 kg
Amperage AC Adapter Input: 0.3 A, AC Adapter Output: 0.84 A
Legal for Trade No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.