missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ AE31E Binocular Microscope
Innovatively designed binocular inverted microscope. Motic™ AE31E Binocular Microscope offers CCSI optics combined with innovative mechanical design, high flexibility, and optical brilliance.
Brand: Motic™ 1100100200584
Additional Details : Weight : 0.01000kg
Description
- Color-corrected infinity optics system
- Auto-off function via IR sensor
- Light memory function
- New generation of LWD plan achromat lenses
- Upgrade options - equipped for fluorescence microscopy
- 12v/100w quartz halogen lamp house
Specifications
Phase shifter Ph1, brightfield, centering telescope, blue filter (OD 45mm), interference filter green (OD 45mm) and frosted glass (OD 45mm) | |
4X, 10X, 20X, 40X | |
5-fold nosepiece, sideways | |
Widefield Eyeglass N-WF10X / 22 mm with diopter adjustment on both eyepieces, incl. eyecups | |
Inverted Binocular Microscope |
ELWD NA 0.30 condenser | |
CCIS Plan Achromatic, CCIS Plan Achromat Phase | |
100/240 V, VDE plug (CE) | |
Halogen |