Learn More
Lovibond™ AF 411 Organo-Phosphonate MINIKIT
Product Code. 10547472
Change view
Click to view available options
Range:
1 to 20 mg/L active O-P
Unit Size:
Each
Product Code. Range Unit Size
10547472 1 to 20 mg/L active O-P Each
1 options
Product Code. 10547472 Supplier Lovibond™ Supplier No. 414110
  • ONLINE EXCLUSIVE PRICE
    178.00 €
    175.60 € / Pack of 1
    Save 2.40 €
    1% Off
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
AF 411 Organo-Phosphonate MINIKIT measures 1-20 mg/L active O-P includes reagents for approximately 30-100 tests uses tablet count method
They are user-friendly and suitable for a wide variety of applications in water analysis.Thanks to various test procedures, the MINIKITs are one thing above all else: a reliable companion.
Various test procedures
For the MINIKITs various test procedures are available:
Choose between manual test procedures, Turbidity or Tablet Count Method or the so-called Speed test.
Intuitive application
Whether professional or layman: with the MINIKIT everyone achieves error-free and reliable results.
Long durability
The reagents used in the MINIKITs have a shelf life of 5 years. And best of all - they are not subject to any transport restrictions.
Directly usable
In addition to the test kit in a sturdy plastic box, the delivery content also includes a measuring beaker, the necessary accessories and tablet reagents for approx. 30 tests.
Specifications
Range 1 to 20 mg/L active O-P
Type Organo-Phosphonate Test Kit
compliance-icons
Product Identifier
  • ORGANO-PHOSPHONATE AF411 MINIKIT .
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: adipic acid