missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B1, crystalline, Thermo Scientific Chemicals

Product Code. 11966201 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 11966201

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aflatoxin B1, crystalline, CAS # 1162-65-8, is a member of a group of mycotoxins presenting potent hepatotoxic and carcinogenic activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Aflatoxin B1 (AFB1) is a member of a group of mycotoxins produced by Aspergillus flavus, and Aspergillus parasiticus showing carcinogenic activities
• AFB1 can induce the transversion of G to T in the third position of codon 249 of the p53 tumor suppressor gene, resulting in a mutation and promoting the carcinogenic process

Applications

• Aflatoxin B1 shows teratogenic activity
• It is a CYP1A2, CYP2A6, CYP2D6, and CYP3A substrate and can be used to study the CYP family in in vitro and in vivo studies.
• It can cause immunosuppression in animal models.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1162-65-8
Molecular Formula C17H12O6
Molecular Weight (g/mol) 312.28
MDL Number MFCD00869647
InChI Key OQIQSTLJSLGHID-UHFFFAOYNA-N
Synonym aflatoxin b1, ccris 12, unii-9n2n2y55mh, afb1, 2,3,6aalpha,9aalpha-tetrahydro-4-methoxycyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 1h,11h-cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione,2,3,6a,9a-tetrahydro-4-methoxy-, 6ar,9as, cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 2,3,6a,9a-tetrahydro-4-methoxy, cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 2,3,6a,9a-tetrahydro-4-methoxy-, 6ar,9as, aflatoxin-b1, aflatoxin b
PubChem CID 186907
ChEBI CHEBI:2504
IUPAC Name 11-methoxy-6,8,19-trioxapentacyclo[10.7.0.0²,⁹.0³,⁷.0¹³,¹⁷]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
SMILES COC1=C2C3=C(C(=O)CC3)C(=O)OC2=C2C3C=COC3OC2=C1

Specifications

Quantity 50 mg
Merck Index 15, 171
Formula Weight 312.27
Chemical Name or Material Aflatoxin B1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.