missing translation for 'onlineSavingsMsg'
Learn More

Aftin-4, MedChemExpress

Product Code. 16473066
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473066 25 mg 25mg
16465306 100 mg 100mg
16434418 10 mg 10mg
16410737 50 mg 50mg
16467137 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473066 Supplier Medchem Express Supplier No. HY111267
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Aftin-4 is an Amyloid-β42 (Aβ42) inducer.

Aftin-4 is an Amyloid-β42 (Aβ42) inducer.

Specifications

Chemical Name or Material Aftin-4
CAS 866893-90-5
Color White
Purity Grade Notes Research
Molecular Formula C20H28N6O
Quantity 25 mg
Solubility Information DMSO : ≥ 100 mg/mL (271.39 mM)
SMILES CC[C@@H](NC1=NC(N(C)CC2=CC=CC=C2)=C3N=CN(C(C)C)C3=N1)CO
Molecular Weight (g/mol) 368.48
Formula Weight 368.48
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.