Learn More
LNI Swissgas AG OFCAS 35 PLUS
Product Code. 17802209 Shop All LNI Swissgas Products
Change view
Click to view available options
Type:
Compressor
Unit Size:
Each
Product Code. Type unitSize
17802209 Compressor Each
1 options
Product Code. 17802209
Brand: LNI Swissgas 6920.96.035
Compressor with dryer integrated.
  • Compressor 35 L/min at 7 bar, 230 V plus with dryer integrated.
Specifications
Type Compressor