missing translation for 'onlineSavingsMsg'
Learn More

Melford Agar Noble

Product Code. 30017999
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 30017999

Brand: Melford A204601000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultra-high purity Agar recommended for use in applications where maximum clarity and purity are essential.

Ultra-high purity Agar recommended for use in applications where maximum clarity and purity are essential. Thorough bleaching and washing procedures are utilized to remove trace amounts of ash and mineral contaminants.

Concentrations of 3-8 g/L obtains a semi-solid gel.

Specifications

Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.