missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agar
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 lb.
5 lb.
Unit Size:
2268g
454g
Description
- Light tan granular powder
- CAS: 9002-18-0
- MDL Number: MFCD00081288
Specifications
Specifications
| Color | Light tan |
| Form | Granulated Powder |
| Product Type | Agar |
| For Use With (Application) | Used as a Gelling Agent in Preparation of Culture Media. |
| Packaging Type | Plastic Bottle |
| Quantity | 1 lb. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction