missing translation for 'onlineSavingsMsg'
Learn More

Agaricinic Acid, TRC

Product Code. 30010397
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010397 1 g 1g
30009793 100 mg 100mg
30006755 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010397 Supplier Toronto Research Chemicals Supplier No. TRCA4256281G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666-99-9
Molecular Formula C22 H40 O7
Molecular Weight (g/mol) 416.55
Synonym Agaric acid (6CI), 2-Hydroxy-1,2,3-nonadecanetricarboxylic acid, Agaricic acid, Agaricin, Agaricinic acid, Laricic acid, NSC 60429, NSC 65690, α-Cetylcitric acid
IUPAC Name 2-hydroxynonadecane-1,2,3-tricarboxylic acid
SMILES CCCCCCCCCCCCCCCCC(C(=O)O)C(O)(CC(=O)O)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H40O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-18(20(25)26)22(29,21(27)28)17-19(23)24/h18,29H,2-17H2,1H3,(H,23,24)(H,25,26)(H,27,28)
Formula Weight 416.2774
Percent Purity >95%
Chemical Name or Material Agaric acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.