missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™

Product Code. 10688973
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10688973 100 g Poly Bottle 100g
10776644 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10688973 Supplier Fisher BioReagents Supplier No. BP1356100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 100 g
DNase DNase free
Electroendosmosis (EEO) 0.15 max.
Gelation Temperature 34.5° to 37.5°C
Gel Strength 1200g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose, Broad Separation Range for DNA/RNA (500bp to 25kb)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.