missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Electrophoresis Grade, MP Biomedicals™

Product Code. 11481682 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481682 500 g 500g
11471682 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481682 Supplier MP Biomedicals Supplier No. 04820723
  • ONLINE EXCLUSIVE PRICE
    1905.00 €
    1804.95 € / 500g
    Save 100.05 €
    5% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Electroendosmosis: 0.13; Gel Strength: ≥600 gm/cm2; Gel Temperature: 36°C; This is a standard agarose with good gel strength and clarity, suitable for all general applications

Chemical Identifiers

CAS 9012-36-6

Specifications

Quantity 500 g
Grade Electrophoresis
Chemical Name or Material Agarose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.