missing translation for 'onlineSavingsMsg'
Learn More

Agarose Gel-Loading Dye, 6X (Contains 15% Ficoll™ 400/Molecular Biology), Fisher BioReagents

Brand:  Fisher Bioreagents BP633-5

Code : 32

Additional Details : CAS Number : 915-67-3 Weight : 0.01080kg

Product Code. 10205023

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Poly Bottle; 5mL

This tracking dye is added to DNA and RNA samples prior to electrophoresis on agarose gels.
Specifications

Specifications

0.3 to 0.6 AU at 252nm, 0.5 to 0.8 AU
Contains 15% Ficoll™ 400
915-67-3,26873-85-8
DNase free
Amaranth (<1.0%), Ficoll™ (15%), Proprietary Component (<1.0%), and Water.
Agarose Gel-Loading Dye, 6X
Liquid
5 mL
Molecular Biology
Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers