missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Ficoll-Based)

Product Code. 15475099
Change view
Click to view available options
Quantity:
10 mL
25 mL
Unit Size:
10mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475099 10 mL 10mL
15485099 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475099 Supplier Thermo Scientific Chemicals Supplier No. J62800.AB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Agarose gel loading dye
Name Note Ficoll based
Physical Form Viscous Liquid
Concentration 6X
Quantity 10 mL
MDL Number MFCD00675058

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.