missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, Genetic Technology Grade

Product Code. 15437479
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437479 25 g 25g
15447479 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437479 Supplier Thermo Scientific Chemicals Supplier No. J66208.14
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 9012-36-6

Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Color White
Quantity 25 g
Sensitivity Hygroscopic; Moisture sensitive
Grade Genetic Technology
Chemical Name or Material Agarose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.