missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, high gelling temperature, Molecular Biology Reagent

Product Code. 15400637
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
15400637 25 g 25g
15410637 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 15400637

Brand: Thermo Scientific Chemicals J66704.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9012-36-6

High gel strength allows gel formation at low concentrations. Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High gel strength allows gel formation at low concentrations
  • Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Color White
Quantity 25 g
Sensitivity Hygroscopic; Moisture sensitive
Chemical Name or Material Agarose, high gelling temperature, Molecular Biology Reagent

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.