missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose medium-low EEO, for electrophoresis, (Mr = 0.12)

Product Code. 10609263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10609263 500 g Glass Bottle 500g
10289590 25 g Glass Bottle 25g
10186980 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10609263 Supplier Thermo Scientific Chemicals Supplier No. 400415000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9012-36-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Fine Crystalline Powder
Melting Point 86.5°C to 89.5°C (1.5% gel)
Color Cream to White
Quantity 500 g
Electroendosmosis (EEO) 0.05 to 0.13
Gelation Temperature 34.5° to 37.5°C (1.5% gel)
Gel Strength >1200g/cm2 (1% gel), >2500g/cm2 (1.5% gel)
Infrared Spectrum Authentic
Packaging Glass Bottle
Grade Electrophoresis
Chemical Name or Material Agarose medium-low EEO
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.