missing translation for 'onlineSavingsMsg'
Learn More

Agarose LE, Molecular Biology Grade, Ultrapure

Product Code. p-7082865 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
Packaging:
Plastic Bottle
missing translation for 'unitSize'
100g
1kg
250g
25g
500g
This item is not returnable. View return policy

Product Code. 15825208

missing translation for 'mfr': Thermo Scientific J32802.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9012-36-6

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Granular Free-flowing Powder
Quantity 25 g
Packaging Plastic Bottle
Grade Molecular Biology
Chemical Name or Material Agarose LE
What are the applications for Agarose LE, Molecular Biology Grade, Ultrapure?

Applications:
1. Routine separation of DNA or RNA fragments.
2. PCR analysis.
3. Blotting applications such as southern blots and northern blots.
4. Protein analysis such as immunodiffusion, rocket assays and immuno electrophoresis.

What are the separation guidelines for Agarose LE, Molecular Biology Grade, Ultrapure?

Range of separation in 1X TAE: 0.6%: 20,000-1000 bp
0.8%: 12,000-500 bp
1.0%: 8000-300 bp
1.2%: 6000-200 bp
1.5%: 3500-100 bp
2.0%: 2000-50 bp

Range of separation in 1X TBE:
0.6%: 15,000-1000 bp
0.8%: 10,000-500 bp
1.0%: 7000-250 bp
1.2%: 5000-200 bp
1.5%: 3000-100 bp
2.0%: 2000-50 bp

How do I prepare an agarose gel?

We recommend the following to prepare an agarose gel:

  1. Weigh out the desired amount of agarose and place it in an Erlenmeyer flask with a measured amount of electrophoresis buffer. Make sure to use the same electrophoresis buffer in the gel as for the running buffer.
  2. Example: For a 0.8% gel, add 0.8 g of agarose and 100 mL of TBE Buffer (1X), to a 200 mL flask. The larger flask insures against the agarose boiling over.
  3. Dissolve the agarose in a boiling water bath or in a revolving-plate microwave oven. All the grains of agarose should be dissolved, and the solution should be clear.
  4. Cool the solution to 60°C (70°C for concentrations 2% or above) and pour immediately.
  5. Allow the gel to set for 30 minutes before using.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.