missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose Low EEO

Product Code. 30017978
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017978 500 g 500g
30017977 50 g 50g
30017975 100 g 100g
30017976 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30017978 Supplier Melford Supplier No. A20080500.0
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Molecular Biology Grade.

Molecular Biology Grade. High purity for preparative and analytical electrophoresis of nucleic acids >1000 bp.

Gelling Temperature:

34.5 -37.5°C

Sulfate:

0.15

EEO:

0.09- 0.13

DNA Binding:

None Detected

DNA and RNase:

None Detected

Specifications

Chemical Name or Material Agarose, Low EEO
CAS 9012-36-6
Quantity 500 g
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.