missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, low gelling temperature, Molecular Biology Reagent

Product Code. 15420557
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420557 100 g 100g
15410557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420557 Supplier Thermo Scientific Chemicals Supplier No. J66319.22
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 9012-36-6

Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility
Soluble in water.

Notes
Gel strength (1.5% w/v):>500g/cm^2; Gelling temperature: <24-28°C; EEO: <0.12
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Melting Point 65°C
Color White
Quantity 100 g
Solubility Information Soluble in water.
Chemical Name or Material Agarose, low gelling temperature, Molecular Biology Reagent

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.