missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Low-Melting, <1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™

Product Code. 10583355
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10583355 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10583355 Supplier Fisher BioReagents Supplier No. BP1360100
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max. | CAS: 9012-36-6 | C12H18O9

Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Agarose
Name Note Low-Melting, <1kb DNA/RNA
Physical Form Powder/Solid
CAS 9012-36-6
Color Cream
Quantity 100 g
DNase DNase free
Electroendosmosis (EEO) 0.15 max.
Gelation Temperature 35°C max.
Gel Strength 500g/cm2min.
Packaging Poly Bottle
Purity Grade Notes DNase- and RNase-Free
Molecular Formula C12H18O9
Formula Weight 306.12
Grade Genetic Analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.