missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Low-Melting, Nucleic Acid Recovery/Molecular Biology Grade), Fisher BioReagents

Product Code. 10377033
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10377033 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10377033 Supplier Fisher BioReagents Supplier No. BP16525
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Physical Form Powder/Solid
CAS 39346-81-1
Color Cream
Quantity 25 g
DNase DNase free
Electroendosmosis (EEO) ≤0.10
Gelation Temperature 26° to 30°C
Gel Strength ≥200g/cm2
Packaging Poly Bottle
Molecular Formula C12H18O9
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose, Low-Melting, Nucleic Acid Recovery
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.