missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose ME

Product Code. p-200087213
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 30017979

Brand: Melford A20085100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For DNA/RNA fragments >1000bp.

For DNA/RNA fragments >1000bp. Intermediate EEO.

Specifications

Chemical Name or Material Agarose ME
CAS 9012-36-6
Quantity 100 g
Grade Molecular Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.