missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Protein Electrophoresis Grade), Fisher BioReagents™

Product Code. 10367033
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10367033 25 g Poly Bottle 25g
10775684 100 g Poly Bottle 100g
10112823 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367033 Supplier Fisher BioReagents Supplier No. BP16425
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9012-36-6 | Molecular Formula: C12H18O9

Agarose (Protein Electrophoresis Grade)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 25 g
DNase DNase free
Electroendosmosis (EEO) 0.1 max.
Gelation Temperature 34° to 45°C
Gel Strength 800g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.