missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Separation ≥500 bp, Genetic Performance Certified™, Ultrapure

Product Code. 15875218 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Plastic Bottle
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15875218 100 g Plastic Bottle 100g
15885218 250 g Plastic Bottle 250g
15895218 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15875218 Supplier Thermo Scientific Supplier No. J75817.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

For PCR, DNA/RNA analytical separation, preparative DNA/RNA recovery, DNA/RNA in-gel recovery, and blotting | CAS: 9012-36-6

Recommended for PCR, DNA/RNA analytical separation, preparative DNA/RNA recovery, DNA/RNA in-gel recovery, and blotting

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 9012-36-6
MDL Number MFCD00081294

Specifikationer

Physical Form Granular
Quantity 100 g
Packaging Plastic Bottle
Grade Molecular Biology
Chemical Name or Material Agarose

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.