missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Standard, Low Electroendosmosis (EEO), Ultrapure Bioreagent, J.T. Baker™

Product Code. 15598664 Shop All J.T.Baker Products
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15598664

Brand: Avantor J.T.Baker A42605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Electrophoresis

Chemical Identifiers

CAS 9012-36-6

Specifications

Quantity 100 g
Packaging Poly Bottle
Grade Ultrapure Bioreagent
Chemical Name or Material Agarose, Standard, Low Electroendosmosis (EEO)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.