missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Agarose Ultra Pure

Product Code. 16052565
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16052565 500 g 500g
16042565 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16052565 Supplier Abnova™ Supplier No. D0021.500g
  • ONLINE EXCLUSIVE PRICE
    1055.00 €
    744.77 € / 500g
    Save 310.23 €
    29% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for everyday analysis of nucleic acids by gele electrophoresis or blotting (Northern or Southern) and is also suitable for protein applications.

No background fluorescence has been detected by ethidium bromide assay. DNase/RNase-free.

Specifications

For Use With (Application) PCR,Electrophoresis
Quantity 500 g
Storage Requirements Store at room temperature Light Sensitive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.