Learn More
Giese™ Agate Mortar with Pestle
Product Code. 11735146
Change view
Click to view available options
Capacity (Metric):
6 mL
13 mL
30 mL
50 mL
80 mL
200 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11735146 30 mL Each
10768031 6 mL Each
10009520 13 mL Each
11745146 50 mL Each
11803360 80 mL Each
11755146 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11735146 Supplier Giese™ Supplier No. 600M080
Spotlight Offers
Use promo code 28038
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Grind up solid chemicals and crush solids into smaller pieces with this agate mortar and pestle. Giese™ Agate Mortar with Pestle features a smooth, dense, and hard surface.
  • Polished agate construction
  • For grinding and crushing
  • Durable
  • Available in different sizes
Specifications
Product Type Mortar and Pestle
Material Agate Stone
Capacity (Metric) 30 mL