missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter™ Agencourt AMPure XP

Product Code. 10136224
Encompass
Change view
Click to view available options
Volume:
5mL
60mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume unitSize
10136224 5mL Each
10453438 60mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10136224 Supplier Beckman Coulter™ Supplier No. A63880

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Trusted and recommended reagent for size selection and cleanup steps within a variety of NGS workflows. Beckman Coulter™ Agencourt AMPure XP removes unwanted contaminants from DNA in a variety of applications including PCR, NGS, cloning, and microarrays.

  • SPRI paramagnetic bead-based chemistry
  • Removes contaminants (dNTPs, salts, primers, primer dimers)
  • High recovery of amplicons (>100 bp) ensures that critical data is not lost
  • Trusted and recommended reagent for a variety of NGS workflows
  • Compatible with manual and automated processing
  • Predictable and consistent size selection
  • Manual cleanups using AMPure XP require less than 30 minutes
  • AMPure XP is automated on all Biomek platforms for higher throughput

Specifications

Composition AMPure XP, Speed Beads (Mag Carboxy)
Content And Storage 2-8°C
Form Liquid
For Use With (Equipment) Automated 96- or 384-well formats, e.g. Biomek platforms
pH 8.0 to 8.4
For Use With (Application) Standard PCR, Real Time PCR (qPCR), Next-Generation Sequencing (NGS)
Sample Type DNA
Volume 5mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.