missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
- Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability
- A ribbon is required to print this material
- Use Part Number M7-R4300 for the BMP71/M710 label printers
Specifications
Specifications
| Certifications/Compliance | UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) for White Only |
| Adhesive Type | Permanent Acrylic |
| Color | White |
| Finish | Matte |
| Format | Die-cut |
| Material | Nylon Cloth |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Component and Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Warehouse Marking, Wire and Cable Labeling |
| For Use With (Equipment) | M710, BMP71 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?