Learn More
Brady™ Aggressive Adhesive Multipurpose Polyester Labels
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady™ handheld label printer.
Brand:  Brady™ 170739
 View more versions of this product
Product Code. 17920770
  • 68.15 € / Pack of 260
Estimated Shipment: 08-12-2026
Add to basket
Description
Description
  • Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Specifications
Specifications
UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863
White
Die-cut
Label
Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;General identification;Inventory and inspection labelling
260
-40°C to 100°C
0.102 mm
1 in.
25.40 mm
25.40 mm
Permanent Acrylic
Gloss
Polyester
Non-sterile
BMP41;BMP51;BMP53;M511;M410;M510
-40°F to 212°F
0.004 in
25.40 mm (W) x 25.40 mm (H)
1 in
1 in
M4-49-422