missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Used for animal immunization, vaccine development and gene therapy, provides an intra-muscular, intra-dermal or intra-tumor electroporation solution to produce maximum transfection efficiency
- Simple user interface
- Pulse Agile Advantage
- Uniform, reliable electroporation
- Simple, Fast Treatment
Specifications
Vaccine Development, Intra-Dermal Gene Delivery, Intra-Muscular Gene Delivery, Intra-Tumor Gene Delivery | |
5 kHz to 1 Hz | |
50 to 1000 V | |
Touchscreen | |
40 cm | |
20 cm | |
50°F to 104°F | |
10°C to 40°C | |
25 lb. | |
12.6 in. | |
Non-sterile | |
Touch Screen Display, Footswitch | |
Needle Array Handle, Miniature Parallel Needle Electrode Array |
Footswitch | |
Electroporation Generator | |
AgilePulse In Vivo IM (Intra-Muscular) Generator Only | |
15.7 in. | |
7.9 in. | |
Generator Only | |
0.200 to 1000 ms | |
0.050 to 10 ms | |
11.3 kg | |
32 cm | |
Electrode Connector Cable | |
Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.