missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ MAX Large Volume Transfection System

Product Code. 15407390
Change view
Click to view available options
Connection Type:
Chamber Cable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
15407390 Chamber Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15407390 Supplier BTX™ Supplier No. 470200N
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AgilePulse™ MAX Large Volume Transfection System provides an advanced electroporation solution for fast, efficient transfection of up to 10 mL of cell suspension.

Includes a user-friendly, programmable waveform generator with patented AgilePulse™ technology, BTX™ patent-pending large-volume electroporation chamber, and proprietary BTXPress™ Cytoporation Medium T, optimized for large volume electroporation. IQ/OQ Now available upon request.

  • Scale-up
    • Transfection protocols readily scale-up from standard laboratory cuvettes to large-volume transfection in the AgilePulse™ MAX system.
  • Maximal Efficiency with Cytoporation Medium
    • BTXPress™ Cytoporation Medium T used with the AgilePulse™ MAX system has been optimized for maximal efficiency with a number of cell lines, including K562, A20, HEK293 and CHO-KI.
    • Compatible with a large range of transfectants including DNA, RNA, siRNA, and olignonucleotides.
    • Can be directly diluted in complete growth medium for post-electroporation cell culture.
  • Simple User Interface
    • All controls are operated with the simple touch screen on the front panel.
    • Data is quickly retrieved by USB key and can be analyzed for detailed pulse characteristics including pulse and pulse current.
  • AgilePulse™ Advantage
    • Transfection efficiency and cell viability are enhanced by specialized, programmable electrical pulse waveforms, particularly important for larger polynucleotide delivery such as DNA plasmids.
    • The patented AgilePulse™ technology combines a unique sequence of short high-intensity pulses to porate cell membranes, followed by long low-intensity pulses to further drive transfectants into cells via electrophoresis, while maintaining cell viability.

Specifications

Format Large volume
For Use With (Application) CRISPR Transfections, Transfect Cells Such as Bone Marrow to Produce or Replace a Missing Protein, Deliver siRNA to Suppress Gene Expression, Deliver Genes for Permanent Gene Correction, Load Cells with a Drug for Drug Delivery, Cancer Immunotherapy, Transfect Eukaryotic Cells for Protein Production in Bioreactors, Large-Scale Production of Replication-Deficient Viruses
For Use With (Equipment) BTXpress Cytoporation Medium T or Cytoporation Medium T4, large volume electroporation chamber
Frequency 5 kHz to 1 Hz
Product Line BTX Electroporation
Product Type Complete Electroporation System
Voltage 50 to 1000 V
Description AgilePulse MAX Large Volume Transfection System, Complete
Display Touchscreen
Depth (English) 15.7 in.
Depth (Metric) 40 cm
Height (English) 7.9 in.
Height (Metric) 20 cm
Includes AgilePulse™ MAX Generator, Large Volume Chamber Stand, Safety Stand, 5 mL Chamber (2 X), Cytoporation Medium T (500 mL) and 4 mm gap Cuvettes
Operating Temperature (English) 50°F to 104°F
Pulse Length Range 0.200 to 1000 ms
Operating Temperature (Metric) 10°C to 40°C
Pulse Width Range 0.050 to 10 ms
Quantity 1 Ea.
Volume (Metric) Up to 10 mL
Weight (English) 25 lb.
Weight (Metric) 11.3 kg
Width (English) 12.6 in.
Width (Metric) 32 cm
Sterility Non-sterile
Connection Type Chamber Cable
Interface Touch Screen Display, Footswitch
Pulse Form Square Wave
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.