missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ Plus Bipolar Transfection System, Complete

Product Code. 16664862
Change view
Click to view available options
Connection Type:
Electrode Connector Cable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
16664862 Electrode Connector Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16664862 Supplier BTX™ Supplier No. 470600N
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Complete AgilePulse™ Plus Bipolar Transfection System can easily perform advanced transfection protocols with multiple groups of pulses, each with separate parameters.

  • Program up to 5 groups of pulses with separately defined pulse number, voltage, pulse length, interval, and polarity - within each group
  • Capacitive Touchscreen Display

Specifications

For Use With (Application) Suspension and adherent cell transfection, Large volume electroporation, CAR-T cell production, CRISPR gene editing, Intradermal, intramuscular and intratumor delivery, Animal immunization and vaccine research
Product Line BTX Electroporation
Product Type Bipolar Transfection System
Voltage 100 to 1200 VDC
Voltage Range (HV Mode) 100 to 1200 VDC
Voltage Range (LV Mode) 100 to 1200 VDC
Description AgilePulse PLUS Bipolar Transfection System, Complete
Display Touchscreen
Field Type Bipolar
Includes AgilePulse PLUS Generator, 5-ft HV Output Cable, F/F Adapters, Extended Capacity Safety Stand, Cytoporation Medium T, 4 mm Gap Cuvettes and Cuvette Rack
Length (English) 14.5 in.
Length (Metric) 5.4 cm
Weight (English) 12.5 lb.
Weight (Metric) 5.7 kg
Width (English) 12 in.
Width (Metric) 30.5 cm
Electrical Requirements 100 to 240 VAC, 50/60 Hz
Sterility Non-sterile
Connection Type Electrode Connector Cable
Interface Touchscreen
Pulse Form Square Wave
High-throughput Compatibility No
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.