missing translation for 'onlineSavingsMsg'
Learn More

Agmatine sulfate, 97%

Product Code. 15491578
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491578 5 g 5g
15401588 250 mg 250mg
15481578 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491578 Supplier Thermo Scientific Alfa Aesar Supplier No. H55363.06
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2482-00-0 | C5H16N4O4S | 228.267 g/mol

Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

Solubility
Soluble in water, nearly insoluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2482-00-0
Molecular Formula C5H16N4O4S
Molecular Weight (g/mol) 228.267
MDL Number MFCD00013109
InChI Key PTAYFGHRDOMJGC-UHFFFAOYSA-N
Synonym agmatine sulfate, 1-4-aminobutyl guanidine sulfate, 1-amino-4-guanidinobutane sulfate salt, 4-aminobutyl guanidinium sulphate, unii-ru0176ql8i, n-4-aminobutyl guanidine sulfate salt, agmatine; sulfuric acid, 4-guanidinobutylamine sulfate salt, guanidine, 4-aminobutyl-, sulfate 1:1
PubChem CID 2794990
IUPAC Name 2-(4-aminobutyl)guanidine;sulfuric acid
SMILES C(CCN=C(N)N)CN.OS(=O)(=O)O

Specifications

Melting Point 231°C to 238°C
Linear Formula H2N(CH2)4NHC(=NH)NH2·H2SO4
Quantity 5 g
Beilstein 3918807
Merck Index 14,188
Solubility Information Soluble in water, nearly insoluble in alcohol
Formula Weight 228.27
Percent Purity 97%
Chemical Name or Material Agmatine sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.