missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agnuside, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Agnuside |
| CAS | 11027-63-7 |
| Molecular Formula | C22 H26 O11 |
| Quantity | 10 mg |
| Synonym | β-D-Glucopyranoside, (1S,4aR,5S,7aS)-1,4a,5,7a-tetrahydro-5-hydroxy-7-[[(4-hydroxybenzoyl)oxy]methyl]cyclopenta[c]pyran-1-yl, Agnuside (6CI,7CI,8CI), β-D-Glucopyranoside, 1,4a,5,7a-tetrahydro-5-hydroxy-7-[[(4-hydroxybenzoyl)oxy]methyl]cyclopenta[c]pyran-1-yl, (1α,4aα,5α,7aα)-, (-)-Buddlejoside A, (1S,4aR,5S,7aS)-1,4a,5,7a-Tetrahydro-5-hydroxy-7-[[(4-hydroxybenzoyl)oxy]methyl]cyclopenta[c]pyran-1-yl β-D-glucopyranoside, Agnoside, Buddlejoside A |
| InChI Formula | InChI=1S/C22H26O11/c23-8-15-17(26)18(27)19(28)22(32-15)33-21-16-11(7-14(25)13(16)5-6-30-21)9-31-20(29)10-1-3-12(24)4-2-10/h1-7,13-19,21-28H,8-9H2/t13-,14+,15+,16+,17+,18-,19+,21-,22-/m0/s1 |
| SMILES | OC[C@H]1O[C@@H](O[C@@H]2OC=C[C@H]3[C@H](O)C=C(COC(=O)c4ccc(O)cc4)[C@@H]23)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | [(1S,4aR,5S,7aS)-5-hydroxy-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-7-yl]methyl 4-hydroxybenzoate |
| Molecular Weight (g/mol) | 466.44 |
| Formula Weight | 466.1475 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction